Learn More
IKA™ Fritte Set
This Fritte Set contains two glass frits for the Pro-Divide reaction cell.
Brand:  IKA™ 0020102633
 View more versions of this product
Product Code. 17573626
  • 70.00 € / Pack of 2
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • This set contains two glass frits for the Pro-Divide reaction cell.
  • The size of each pore falls between these ranges:
    • 1 μm set: 1 and 1.6 μm
    • 5 μm set: 4 and 5.5 μm
    • 10 μm set: 10 and 16 μm
Specifications
Specifications
Fritte Set
5 μm
Pro-Divide Reaction Cell