missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ Fritte Set
This Fritte Set contains two glass frits for the Pro-Divide reaction cell.
70.00 € - 149.00 €
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17563626
|
IKA™
0020102632 |
1 μm |
149.00 €
Pack of 2 |
Estimated Shipment: 13-03-2025
Log in to see stock. |
|||||
17573626
|
IKA™
0020102633 |
5 μm |
70.00 €
Pack of 2 |
Estimated Shipment: 13-03-2025
Log in to see stock. |
|||||
17583626
|
IKA™
0020102634 |
10 μm |
79.00 €
Pack of 2 |
Estimated Shipment: 13-03-2025
Log in to see stock. |
|||||
Description
- This set contains two glass frits for the Pro-Divide reaction cell.
- The size of each pore falls between these ranges:
- 1 μm set: 1 and 1.6 μm
- 5 μm set: 4 and 5.5 μm
- 10 μm set: 10 and 16 μm
Specifications
Fritte Set | |
Pro-Divide Reaction Cell |