Learn More
Thermo Scientific Chemicals FSB Buffer
For frozen storage of competent cells
Brand:  Thermo Scientific Chemicals J60262.AK
 View more versions of this product
Product Code. 15433059
  • 149.75 € / 250mL
Explore more special offers
Description
Description
For frozen storage of competent cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Colorless
Frozen storage buffer
Liquid
Buffer
250 mL
2°C to 8°C
For frozen storage of competent cells