missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals FSB Buffer
For frozen storage of competent cells
87.60 € - 149.75 €
Specifications
Color | Colorless |
---|---|
Synonym | Frozen storage buffer |
Content And Storage | 2°C to 8°C |
Form | Liquid |
For Use With (Application) | For frozen storage of competent cells |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15423059
|
Thermo Scientific Chemicals
J60262.=E |
125 mL |
87.60 €
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15433059
|
Thermo Scientific Chemicals
J60262.AK |
250 mL |
149.75 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For frozen storage of competent cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Colorless | |
2°C to 8°C | |
For frozen storage of competent cells |
Frozen storage buffer | |
Liquid | |
Buffer |
Safety and Handling
Recommended Storage : Keep cold
RUO – Research Use Only