Learn More
JULABO™ Immersion Cooler, FT Series
Primarily used for countercooling open bath circulators to ambient temperature or below as well as for rapidly cooling liquids. JULABO™ immersion coolers have a small footprint and represent an economic solution preventing tap water usage for cooling.
5375.00 € - 10885.00 €
Specifications
Description Immersion Cooler FT
Voltage 115/230 V
View More Specs
Products 2
Product Code Brand Temperature Range Includes Price Quantity & Availability  
Product Code Brand Temperature Range Includes Price Quantity & Availability  
10264011
View Documents
JULABO™
9650840
-40°C to +30°C Immersion probe (L x dia.) 12 x 5 cm
5375.00 €
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
11994818
View Documents
JULABO™
9650890
-90°C to +30°C Flexible immersion probe (L x dia.) 65 x 1.5 cm
10885.00 €
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
Description
Description
  • Rapid cooling of liquids down to low temperatures
  • Counter-cooling of liquids in combination with heating circulators
  • Environmentally friendly by saving precious tap water
  • User-friendly operation and handling
  • Compact design, small footprint
  • Dry ice substitution
Specifications
Specifications
Immersion Cooler FT
115/230 V