missing translation for 'onlineSavingsMsg'
Learn More

MULTIROIR Full Eurobox Container

Full Eurobox container with solid bottoms and solid walls, ideal for the storage and transport of goods

Brand:  MULTIROIR BA12019

 View more versions of this product

Product Code. 17276900

This item is not returnable. View return policy

Description

Description

  • Full Eurobox container
  • The best quality / price ratio on the market
  • Solid bottoms and solid walls
  • Ideal for the storage and transport of goods
  • Stackable and high load capacity
  • Temperature resistant from -20 ° C to + 110 ° C
Specifications

Specifications

Storage Bin
Red
600 x 400 x 100 mm
Polypropylene
Rectangular
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MULTIROIR Full Eurobox Container > 600 x 400 x 100 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.