Learn More
FUT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18197-1-AP-150UL
 View more versions of this product
Product Code. 16848254
  • 410.00 € / 150µL
Explore more special offers
Description
Description
May catalyze alpha-1,3 glycosidic linkages involved in the expression of sialyl Lewis X antigens.
Specifications
Specifications
FUT7
Polyclonal
Unconjugated
FUT7
Alpha (1,3) fucosyltransferase, Fuc TVII, Fucosyltransferase 7, Fucosyltransferase VII, FucT VII, FUT7, Selectin ligand synthase
Rabbit
Antigen Affinity Chromatography
RUO
14347, 2529, 296564
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q11130, Q11131
Fut7
FUT7 Fusion Protein Ag12475
150 μL
Primary
Human, Mouse, Rat