missing translation for 'onlineSavingsMsg'
Learn More

FUT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen FUT7
Concentration 0.47 mg/mL
Applications Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16858254
View Documents
Proteintech
18197-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16848254
View Documents
Proteintech
18197-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

May catalyze alpha-1,3 glycosidic linkages involved in the expression of sialyl Lewis X antigens.
Specifications

Specifications

FUT7
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Unconjugated
Rabbit
Human, Rat, Mouse
Q11130, Q11131
14347, 2529, 296564
FUT7 Fusion Protein Ag12475
Primary
-20°C
FUT7
0.47 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Alpha (1,3) fucosyltransferase, Fuc TVII, Fucosyltransferase 7, Fucosyltransferase VII, FucT VII, FUT7, Selectin ligand synthase
Fut7
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.