Learn More
Syngene G:Box Mini XRQ Compact Imaging System

G:Box Mini XRQ is a compact system packed full of features to seamlessly integrate into your research. Designed to adapt to multi-user labs and imaging across the full spectrum.
Brand: Syngene GBOX-MINI-XRQ
Description
G:Box mini XRQ is a compact system that may be small but is packed full of features to seamlessly integrate the G:Box mini system into your research.
This imaging system is designed to adapt to the diverse needs of multi-user labs. It's not just for blots and gels with HILED modules, it provides robust excitation for quantitative fluorescence imaging. You can image across the full spectrum UV, Blue, Green, Red, Far Red and IR, giving you the flexibility to meet all of your imaging needs. This adaptability empowers you to explore a wide range of scientific applications.
A sensitive system with a wide dynamic range, detects both weak and strong bands on the same blot without compromising quality using intuitive GeneSys image capture software. A large aperture lens combined with a high-resolution camera allows for unmatched image quality and sensitivity.
- Capable of imaging many applications DNA/Protein gels, chemiluminescence, stain free gels, colored multiplex fluorescence blots, in-vivo applications (additional accessories may be needed).
- Peltier cooled camera with 4 MP image resolution capable of producing 16-bit images (with 65535 grayscales).
- System has an F1.2 lens with motorized iris, focus and zoom (with feedback) for short acquisitions.
- System comes with 7 position filter wheel. Epi white light and option of HILED's which gives capability for fluorescence imaging for red, green, blue, far red and IR imaging (additional accessories will need to be purchased).
- Maximum viewing area 18 x 14 cm.
- GeneSys image capture software with its user- friendly interface produces publication quality images.
- GeneTools software provides the user with rapid quantification of samples including molecular weight, colony counting, protein normalisation and multiplex fluorescence analysis.
- Software can be used in 21 CFR part 11 and GLP environments.
- IQOQPQ documentation available on request.
Specifications
None | |
Gel Imaging | |
Gel Imaging System |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.