missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

94.07 € - 656.27 €

Chemical Identifiers

CAS 13450-84-5
Molecular Formula Cl3GdH12O6
Molecular Weight (g/mol) 371.69
MDL Number MFCD00149732
InChI Key PNYPSKHTTCTAMD-UHFFFAOYSA-K
Synonym gadolinium chloride hexahydrate, gadolinium trichloride hexahydrate, unii-261ex557hi, gadolinium chloride, hydrate 1:6, gadolinium chloride gdcl3 , hexahydrate, gadolinium chloride gdcl3 , hexahydrate 8ci,9ci, gadolinium 3+ trichloride hexahydrate, gadolinium iii chloridehexahydrate, gadolinium chloride, hexahydrate,, gadolinium iii chloride hexahydrate, reacton reo
PubChem CID 197915
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11391009
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011287.06
5 g
94.07 €
5g
Please to purchase this item. Need a web account? Register with us today!
11301019
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011287.14
25 g
222.11 €
25g
Please to purchase this item. Need a web account? Register with us today!
11311019
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011287.22
100 g
656.27 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gadolinium(III) chloride hexahydrate is employed in synthesis and NMR studies of the paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes with possible use as contrast agents for magnetic resonance imaging..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13450-84-5
371.69
PNYPSKHTTCTAMD-UHFFFAOYSA-K
197915
Cl3GdH12O6
MFCD00149732
gadolinium chloride hexahydrate, gadolinium trichloride hexahydrate, unii-261ex557hi, gadolinium chloride, hydrate 1:6, gadolinium chloride gdcl3 , hexahydrate, gadolinium chloride gdcl3 , hexahydrate 8ci,9ci, gadolinium 3+ trichloride hexahydrate, gadolinium iii chloridehexahydrate, gadolinium chloride, hexahydrate,, gadolinium iii chloride hexahydrate, reacton reo
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]

Specifications

13450-84-5
Crystalline Aggregates
MFCD00149732
gadolinium chloride hexahydrate, gadolinium trichloride hexahydrate, unii-261ex557hi, gadolinium chloride, hydrate 1:6, gadolinium chloride gdcl3 , hexahydrate, gadolinium chloride gdcl3 , hexahydrate 8ci,9ci, gadolinium 3+ trichloride hexahydrate, gadolinium iii chloridehexahydrate, gadolinium chloride, hexahydrate,, gadolinium iii chloride hexahydrate, reacton reo
PNYPSKHTTCTAMD-UHFFFAOYSA-K
gadolinium(3+);trichloride;hexahydrate
197915
99.99%
Characteristic
2.424 g/mL
Gadolinium(III) chloride hexahydrate
White
Cl3GdH12O6
14,4326
soluble in water.
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]
371.69
371.70 (263.61 Anhydrous)
Reacton™
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'einecsNumber' : 233-386-6

missing translation for 'rtecsNumber' : LW4050000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • Gadolinium(III) chloride hexahydrate, REacton

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Gadolinium chloride(GdCl3), hexahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only