missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) nitrate hydrate, 99.5% (REO), Thermo Scientific Chemicals

Chemical Identifiers

CAS 94219-55-3
Molecular Formula GdN3O9
Molecular Weight (g/mol) 343.26
MDL Number MFCD00011023
InChI Key MWFSXYMZCVAQCC-UHFFFAOYSA-N
IUPAC Name gadolinium(3+) trinitrate
SMILES [Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11397647
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012917.14
25 g N/A
N/A
11387647
missing translation for 'viewDocuments' Détails de la promotion
Thermo Scientific Alfa Aesar
012917.22
100 g N/A
N/A
Description

Description

It is applied as an oxidizing agent and the precursor material in the preparation of gadolinium compounds. It is extensively employed in the production of specialized glasses and ceramics. It is also employed in heavy water nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécification

Identifiants chimiques

94219-55-3
343.26
MWFSXYMZCVAQCC-UHFFFAOYSA-N
[Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
GdN3O9
MFCD00011023
gadolinium(3+) trinitrate

Spécification

92°C
White
GdN3O9
UN1477
Soluble in water.
[Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
343.26
99.9%
2.406 g/mL
Gadolinium(III) nitrate hydrate
94219-55-3
Crystalline
MFCD00011023
14,4326
MWFSXYMZCVAQCC-UHFFFAOYSA-N
gadolinium(3+) trinitrate
343.26 (Anhydrous)
Hygroscopic
(REO)
Vidéos
Sécurité et traitement

Sécurité et traitement

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

missing translation for 'rtecsNumber' : LW4550000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

FDS
Documents

Documents

Certificats

RUO – Research Use Only