Learn More
Fisherbrand™ Gamma Radiation Shields
Gamma radiation shields with fixed 15 degree angles and a curved base of various dimensions, are ideal for protection from Gamma radiation.
Brand:  Fisherbrand™ CSR-S2TG
 View more versions of this product
Product Code. 12357187
  • 2765.00 € / Each
Estimated Shipment: 28-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Radiation shielding Cleaver Scientific for protection from Gamma radiation, shield, fixed 15 degree angle, curved base, dimensions 530h x 350w x 150d mm
  • Provides clear, non-distorted vision, reducing light images and shadows, especially when the user is in a sitting position
  • Supplied in small, medium and large sizes and with curved base 15cm-deep for use with safety trays or flat 30x30cm base for under-the-bench protection
  • A range of angles offers increased maneuverability, while clear optical acrylic aids visualization
  • Available in two sizes, with either a flat base to provide bench protection, or a curved base which fits conveniently over safety trays
Specifications
Specifications
15° Angle
Gamma Radiation Shield
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Gamma Radiation Shields > Angle: Large Fixed 15° Height: 53cm