Learn More
Fisherbrand™ Gamma Radiation Shields
Gamma radiation shields with fixed 15 degree angles and a curved base of various dimensions, are ideal for protection from Gamma radiation.
2435.00 € - 3275.00 €
Specifications
Product Type Gamma Radiation Shield
View More Specs
Products 3
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
12357187
missing translation for 'viewDocuments'
Fisherbrand™
CSR-S2TG
15° Angle
2765.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
12337187
missing translation for 'viewDocuments'
Fisherbrand™
CSR-S2OTG
45° Angle
3275.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
12337177
missing translation for 'viewDocuments'
Fisherbrand™
CSR-S1OG
Small Fixed 45° Angle
2435.00 €
Each
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
Description
Description
  • Radiation shielding Cleaver Scientific for protection from Gamma radiation, shield, fixed 15 degree angle, curved base, dimensions 530h x 350w x 150d mm
  • Provides clear, non-distorted vision, reducing light images and shadows, especially when the user is in a sitting position
  • Available in two sizes, with either a flat base to provide bench protection, or a curved base which fits conveniently over safety trays
Specifications
Specifications
Gamma Radiation Shield
Videos
SDS
Documents
Documents
Certificates
Product Content Correction
Product Title
Fisherbrand™ Gamma Radiation Shields