Learn More
Cytiva GammaBind™ Plus Sepharose™ Medium
Engineered for better binding to rat and mouse IgG subclasses
Brand:  Cytiva 17-0886-04
 View more versions of this product
Product Code. 11504955
  • 22200.00 € / 500mL
Explore more special offers
Description
Description
  • Recombinant Protein G type 3 coupled to Sepharose CL-6B for easy binding and fast separation
  • More enhanced binding capabilities than GammaBind G at both laboratory and process scale
  • Yields up to 18mg/mL mouse IgG
  • Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GammaBind Plus Sepharose
90 μm
500 mL
Recombinant GammaBind G type 3 lacking albumin-binding region
2°C to 8°C, 20% Ethanol
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva GammaBind™ Plus Sepharose™ Medium > 500mL