Learn More
Thermo Scientific™ Gas Chromatography Tubing
Find the copper and stainless steel tubing in a variety of sizes for plumbing your gas chromatography (GC) system.
Brand:  Thermo Scientific™ 60181-638
 View more versions of this product
Product Code. 15204613
  • 150.27 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Find copper and stainless steel Thermo Scientific™ Gas Chromatography Tubing in a variety of sizes for plumbing your gas chromatography (GC) system.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gas Chromatography Tubing
25 ft. Length
1/8 in.
Stainless Steel
0.085 in.
Gas Chromatography Systems
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.