missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gas Chromatography Tubing

Find the copper and stainless steel tubing in a variety of sizes for plumbing your gas chromatography (GC) system.

98.15 € - 150.27 €

Specifications

Type Gas Chromatography Tubing
For Use With (Equipment) Gas Chromatography Systems
View More Specs

Products 8
Product Code Brand Material Diameter (English) Inner Diameter (English) Outer Description Price Quantity & Availability  
Product Code Brand Material Diameter (English) Inner Diameter (English) Outer Description Price Quantity & Availability  
15244603
View Documents
Thermo Scientific™
60181-632
Copper 0.065 in. 1/8 in. 50 ft. Length
119.00 €
Each
In Stock
to see stock.
Add to basket
 
15254603
View Documents
Thermo Scientific™
60181-633
Copper 0.19 in. 1/4 in. 50 ft. Length This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15264603
View Documents
Thermo Scientific™
60181-634
Stainless Steel 0.01 in. 1/16 in. 25 ft. Length This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15274603
View Documents
Thermo Scientific™
60181-635
Stainless Steel 0.02 in. 1/16 in. 25 ft. Length This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15284603
View Documents
Thermo Scientific™
60181-636
Stainless Steel 0.03 in. 1/16 in. 25 ft. Length This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15294603
View Documents
Thermo Scientific™
60181-637
Stainless Steel 0.04 in. 1/16 in. 25 ft. Length This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15204613
View Documents
Thermo Scientific™
60181-638
Stainless Steel 0.085 in. 1/8 in. 25 ft. Length
150.27 €
Each
Estimated Shipment: 18-12-2024
to see stock.
Add to basket
 
15214613
View Documents
Thermo Scientific™
60181-639
Stainless Steel 0.21 in. 1/4 in. 25 ft. Length This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Find copper and stainless steel Thermo Scientific™ Gas Chromatography Tubing in a variety of sizes for plumbing your gas chromatography (GC) system.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications

Specifications

Gas Chromatography Tubing
Gas Chromatography Systems
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.