missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Gas and Vapour Filters
3M™ Gas and Vapour Filters; reusable masks with a bayonet connection and are designed to optimise your field of vision. Our filters offer lightweight and well-balanced breathing protection for hazardous environments.
Brand: 3M™ 7000059883
155.71 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Our P3 R particulate filters have more surface area than many standard filters and offer lightweight and well-balanced breathing protection for hazardous environments. For P3 R breathing protection against solid and liquid particles, and hydrogen fluoride gas up to 30 ppm (or up to 10 x OEL), use 3M™ Particulate Filters 6038. They also provide additional protection against nuisance-level organic vapours and acid gases below OEL that are encapsulated in a plastic case for rough applications.Our particulate filters offer more surface area than many standard filters. The surface area is 1.8 x larger than 3M™ Particulate Filter 2000 Series filters and can potentially increase service life in some applications. They offer low breathing resistance and good balance when connected to your mask and are designed to optimise your field of vision. They can be clicked into place quickly and easily, and they can be used with any 3M™ Reusable Masks that feature a bayonet-style connection. The hard case protects against sparks and high heat, and it’s ideal for harsh environments where easy decontamination and protection is needed.
Specifications
EN143 : 2000 | |
Protects against solids and liquid particles P3R, HF up to 10 x TLV additionally against organic vapour and acid gases below OEL in a stable plastic cage for rough applications. | |
Particulate Filter | |
Suitable for 3M™ 6000 and 3M™ 7000 Series Half Face and Full Face Masks | |
10/Pk. |
Bayonet | |
P3R | |
Grinding, Sanding, Polishing, Food manufacturing, Construction/quarrying, Woodworking, Pharmaceutical/powdered chemicals, Water blasting, Fiberglass installation, Asbestos related tasks | |
6038 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Gas and Vapour Filters > Model: 6038
Spot an opportunity for improvement?Share a Content Correction