Learn More
3M™ Gas and Vapour Filters
3M™ Gas and Vapour Filters; reusable masks with a bayonet connection and are designed to optimise your field of vision. Our filters offer lightweight and well-balanced breathing protection for hazardous environments.
77.63 € - 560.00 €
Specifications
Connection Type Bayonet
View More Specs
Products 8
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
12352339
View Documents
3M™
7000034747
6059
205.00 €
4 pairs
In Stock
to see stock.
Edge
Add to basket
15749007
View Documents
3M™
7000030099
6099 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15729067
View Documents
3M™
7100006000
6057
93.55 €
4 pairs
In Stock
to see stock.
Edge
Add to basket
15789007
View Documents
3M™
7000034750
6054
560.00 €
32 pairs
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
16802016
View Documents
3M™
7000034749
6055
82.20 €
4 pairs
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
15769007
View Documents
3M™
7000034746
6075
81.10 €
4 pairs
In Stock
to see stock.
Edge
Add to basket
15799007
View Documents
3M™
7000034751
6051
163.00 €
4 pairs
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
15759027
View Documents
3M™
7000059883
6038
229.00 €
10 pairs
In Stock
to see stock.
Edge
Add to basket
Description
Description
Colour-coded labels according to the EN14387 classification system to show the type of protection they provide.These lightweight filters combine protection with comfort, and they’re compatible with our range of half and full-face masks for safe working in hazardous environments. They offer low breathing resistance and good balance when connected to your mask, and they’re designed to optimise your field of vision.They can be clicked into place quickly and easily, and they can be used with any 3M™ Reusable Masks that feature a bayonet-style connection. They can also be used in combination with 3M™ Particulate Filters and 3M™ 501 Particle Filter Retainers.
Specifications
Specifications
Bayonet
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
3M™ Gas and Vapour Filters