missing translation for 'onlineSavingsMsg'
Learn More

GATA3 Mouse anti-Human, Mouse, Clone: 1B11A8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66400-1-IG-20UL

 View more versions of this product

Product Code. 16838245

  • 145.00 € / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Trans-acting T-cell specific transcription factor GATA-3 is a member of GATA family of transcription factors that regulates development of multiple tissues. It is an important transcription factor in regulating human Th2 cell differentiation in vivo.
Specifications

Specifications

GATA3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P23771, P23772
Gata3
GATA3 Fusion Protein Ag0174
20 μL
Primary
Mouse, Human
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
1B11A8
Unconjugated
GATA3
GATA binding factor 3, GATA binding protein 3, GATA3, HDR
Mouse
Protein A
RUO
14462, 2625
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.