missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA3 Mouse anti-Human, Mouse, Clone: 1B11A8, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | GATA3 |
---|---|
Clone | 1B11A8 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838245
|
Proteintech
66400-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16828245
|
Proteintech
66400-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Trans-acting T-cell specific transcription factor GATA-3 is a member of GATA family of transcription factors that regulates development of multiple tissues. It is an important transcription factor in regulating human Th2 cell differentiation in vivo.Specifications
GATA3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GATA binding factor 3, GATA binding protein 3, GATA3, HDR | |
Gata3 | |
IgG2a | |
Protein A | |
Antibody |
1B11A8 | |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Mouse, Human | |
P23771, P23772 | |
14462, 2625 | |
GATA3 Fusion Protein Ag0174 | |
Primary | |
-20°C | |
GATA3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title