Learn More
Lenz Laborglasintrumente™ Gay Lussac Pycnometers Unadjusted
Pycnometers acc. to Gay Lussac, unadjusted
Brand:  Lenz Laborglasintrumente™ 05630022
 View more versions of this product
Product Code. 10788081
  • 44.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Gay Lussac pycnometers acc. to DIN ISO 3507 and in addition to standard
  • Unadjusted
  • Stopper NS 10/19 with capillary
  • Stopper top ground and polished
  • Imprint of nominal capacity at the bottom
  • Material: borosilicate glass 3.3
Specifications
Specifications
1
25 mL
Borosilicate Glass (3.3)