missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Gay Lussac Pycnometers Unadjusted

Pycnometers acc. to Gay Lussac, unadjusted

44.00 € - 45.00 €

Specifications

Quantity 1
Material Borosilicate Glass (3.3)
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10788081
View Documents
Lenz Laborglasintrumente™
05630022
25 mL
44.00 €
Each
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
 
10000281
View Documents
Lenz Laborglasintrumente™
05630028
50 mL
45.00 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
Description

Description

  • Gay Lussac pycnometers acc. to DIN ISO 3507 and in addition to standard
  • Unadjusted
  • Stopper NS 10/19 with capillary
  • Stopper top ground and polished
  • Imprint of nominal capacity at the bottom
  • Material: borosilicate glass 3.3
Specifications

Specifications

1
Borosilicate Glass (3.3)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasintrumente™ Gay Lussac Pycnometers Unadjusted

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.