missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ GC Inlet Liner Splitless Straight-Through Liner For Perkinelmer Autosystem & Clarus 500 2mm I.D.
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 09210301
This item is not returnable.
View return policy
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with PerkinElmer instruments
Specifications
2 mm | |
92 mm | |
Splitless, Straight-through Inlet Liner | |
6.2 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trajan Scientific and Medical™ GC Inlet Liner Splitless Straight-Through Liner For Perkinelmer Autosystem & Clarus 500 2mm I.D. >
Spot an opportunity for improvement?Share a Content Correction