Learn More
Invitrogen™ Human GDF15 ELISA Kit
ELISA
Brand: Invitrogen™ EHGDF15X5
2451.48 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
The Human GDF15 ELISA quantitates Hu GDF15 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu GDF15. Principle of the method The Human GDF15 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
GDF-15 belongs to the TGF-beta cytokine family, whose members play an important role during prenatal development and postnatal growth, and the remodeling and maintenance of a variety of tissues and organs. GDF-15 is expressed predominantly in the placenta and, to a much lesser extent, in various other tissues. The presence of GDF-15 in amniotic fluid and its elevated levels in the sera of pregnant women suggest GDF-15's involvement in gestation and embryonic development. GDF-15 generally exerts tumor suppressive activities and is one of the predominant factors produced and secreted in response to activation of the p53 pathway. Interestingly, the serum level of GDF-15 is positively correlated with neoplastic progression of several tumor types, including certain colorectal, pancreatic, and prostate cancers.
Specifications
Q99988 | |
2 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA | |
Plasma, Serum, Supernatant | |
5 x 96 Assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
9518 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
5 x 96 Tests | |
Signal Transduction | |
2°C to 8°C | |
GDF15 | |
4 hrs 45 mins |
1.1 to 800 pg/mL | |
2 pg/mL to 800 pg/mL | |
HRP | |
Growth Factors & Receptors | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA- 78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL- 11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF- 7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
Human | |
Colorimetric Microplate Reader | |
GDF-15,MIC-1,MIC1,NAG-1,PDF,PLAB,PTGFB | |
<10 | |
HRP | |
RUO | |
Plasma 33 μL, Serum 33 μL, Supernatant 100 μL | |
GDF-15, MIC-1, MIC1, NAG-1, PDF, PLAB, PTGFB | |
1 hrs 20 mins |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.