Learn More
Cytiva Gel Filtration Calibration Kits
Calibrate gel filtration columns simply, reliably. Cytiva Life Sciences™ Gel Filtration Calibration Kits contain a well-defined series of protein standards.
Brand:  Cytiva 28-4038-41
618.45 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
 View more versions of this product
Product Code. 10734557
  • 670.00 € / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Optimized range of proteins suited to high-resolution media and prepacked columns
  • Each kit contains five vials of lyophilized proteins
  • Blue Dextran 2000 included to determine the void fraction of the column
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Each
Calibration Kit
10mg Aprotinin (6500 Mr), 50mg Ribonuclease A (13,700 Mr), 15mg Carbonic anhydrase (29,000 Mr), 50mg Ovalbumin (43,000 Mr), 50mg Conalbumin (75,000 Mr), 50mg Blue Dextran (2,000,000 Mr)
2° to 8°C
High-resolution media and prepacked column
Calibration of high-resolution gel filtration media and column such as Superdex™, Superose and Sephacryl
6500 to 75,000
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Supplemental information
  • MIXTURE LIST-Contains : Trypsin inhibitor, pancreatic basic