Learn More
Gibco™ Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix
A soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors. It is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation.
Brand:  Gibco™ A1413201
51.54 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
 View more versions of this product
Product Code. 11692139
  • 54.25 € / 1mL
Edge
Explore more special offers
Description
Description
  • The major components of Geltrex™ matrix include laminin, collagen IV, entactin, and heparin sulfate proteoglycans
  • It is ideal for the promotion and maintenance of differentiated phenotypes in a variety of cell cultures including primary epithelial cells, endothelial cells, smooth muscle cells, and human induced pluripotent stem cells (iPSC)
  • Geltrex™ matrix offers, on average, a 2-fold reduction in several growth factors
  • As a result, analysis of specific growth factor effects in applications requiring basement membrane extract can be carried out in a controlled and reproducible format
  • Minimal protein concentration variation from lot-to-lot (∼15 mg/mL)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matric comes as a sterile protein solution in 1 mL vials.
Store at -20 to -80°C.
Geltrex™ Matrix is stable for 18 months from date of manufacture when properly stored.
Reduced Growth Factor
Adherent Cell Culture
Basement Membrane Matrix
Sterile
Low
Gibco™
1 mL
Mouse
Variety of cultured cells, including primary epithelial cells, endothelial cells, smooth muscle cells, and human induced pluripotent stem cells (iPSC)
12 to 18 mg/mL
Frozen
Serum-free
No Phenol Red
Tube(s)
Cell Culture Grade
Dry Ice