missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix
A soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors. It is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation.
57.50 € - 265.00 €
Specifications
Cell Type | Variety of cultured cells, including primary epithelial cells, endothelial cells, smooth muscle cells, and human induced pluripotent stem cells (iPSC) |
---|---|
Classification | Reduced Growth Factor |
Concentration | 12 to 18 mg/mL |
Culture Type | Adherent Cell Culture |
Form | Frozen |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12053569
|
Gibco™
A1413202 |
5 mL |
265.00 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11692139
|
Gibco™
A1413201 |
1 mL |
57.50 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The major components of Geltrex™ matrix include laminin, collagen IV, entactin, and heparin sulfate proteoglycans
- It is ideal for the promotion and maintenance of differentiated phenotypes in a variety of cell cultures including primary epithelial cells, endothelial cells, smooth muscle cells, and human induced pluripotent stem cells (iPSC)
- Geltrex™ matrix offers, on average, a 2-fold reduction in several growth factors
- As a result, analysis of specific growth factor effects in applications requiring basement membrane extract can be carried out in a controlled and reproducible format
- Minimal protein concentration variation from lot-to-lot (∼15 mg/mL)
Specifications
Variety of cultured cells, including primary epithelial cells, endothelial cells, smooth muscle cells, and human induced pluripotent stem cells (iPSC) | |
12 to 18 mg/mL | |
Frozen | |
Serum-free | |
No Phenol Red | |
Tube(s) | |
Cell Culture Grade | |
Mouse |
Reduced Growth Factor | |
Adherent Cell Culture | |
Basement Membrane Matrix | |
Sterile | |
Low | |
Gibco™ | |
Dry Ice |
Safety and Handling
missing translation for 'shelfLife' : 36 months
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 2-Bromo-5-chlorotoluene
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 2-Bromo-5-chlorotoluene
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title