Learn More
Agilent BioTek Gen5 Data Collection and Analysis Software
Gen5 Data Collection and Analysis Software has a contemporary interface which provides easy access to all major functions of the software including reading parameters, plate layout, data reduction, online help, wizards, tutorials and sample protocols.
Brand:  Agilent BioTek GEN5AL2
 View more versions of this product
Product Code. 15649408
  • 304.00 € / Each
Explore more special offers
Description
Description
  • Quick launch buttons on the welcome screen provide links to recently used files, pre-programmed protocols and other key functions such as Quick Read and System Test
  • StepWise tools allow limitless combinations of procedural steps and data reduction criteria for a wide variety of applications in a logical and clearly defined sequence that is easily viewed on the screen
  • Analyze data on a single plate or across multiple plates in an experiment
  • Report and PowerExport builders are fast and flexible output tools to produce publication-ready reports using data matrices, statistics tables, protocols and plate information
  • For compliance to 21 CFR Part 11, Gen5 Secure provides system, data and protocol protection
  • Multiple user definitions and permission levels make it easy to comply with laboratory policies
System Requirements
  • Windows XP or Windows 2000 (Professional Editions)
  • PentiumIII-Class PC (or compatible) processor (500MHz or higher)
  • 512 MB RAM or higher
  • 2 GB hard drive space or higher
  • XGA resolution at 1024 x 768 or higher
  • CD-ROM readable drive
  • Keyboard
  • Mouse
  • Microsoft Internet Explorer v 5.0 or higher (for online help)
  • Microsoft Excel 2000 or later (for PowerExport feature)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gen5 AL2 Data Analysis Software
Imaging Multi-Mode Readers
Data Collection and Analysis Software
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Agilent BioTek Gen5 Data Collection and Analysis Software > Gen5 AL2