Learn More
Brannan™ Lab Thermometer with 3-points UKAS Calibration
General purpose laboratory thermometer complete with calibration certificate.
Brand:  Brannan™ 46/456/0 + 49/503/0
 View more versions of this product
Product Code. 17823088
  • 293.00 € / Each
Explore more special offers
Description
Description
  • Lab thermometer
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications
Specifications
-60°C, 0°C, 40°C
Total Immersion
Glass
122°F
-148°F
-148°F to +122°F
± 2.0°C at 35°C
1.2 cm
ISO 17025 UKAS
30.5 cm
Red Spirit
50°C
-100°C
-100°C to +50°C
1°C