Learn More
Brannan™ Lab Thermometer with 3-points UKAS Calibration
General purpose laboratory thermometer complete with calibration certificate.
183.00 € - 293.00 €
Specifications
Certifications/Compliance | ISO 17025 UKAS |
---|---|
Material | Glass |
Width (Metric) | 1.2 cm |
Product Code | Brand | Temperature Range (Metric) | Thermometer Fill | Immersion Type | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Thermometer Fill | Immersion Type | Length (Metric) | Price | Quantity & Availability | |||
17842978
|
Brannan™
44/403/3 + 49/503/0 |
-10°C to +110°C | Green Spirit | Partial Immersion | 30.5 cm |
184.00 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||
17893018
|
Brannan™
44/450/0 + 49/503/0 |
-10°C to +50°C | Red Spirit | Total Immersion | 15 cm |
185.00 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||
17863008
|
Brannan™
44/002/7 + 49/503/0 |
-18°C to +110°C | Spirit | Partial Immersion | 30.5 cm |
186.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17823298
|
Brannan™
44/002/8 + 49/503/0 |
-20°C to +110°C | Spirit | Partial Immersion | 30.5 cm |
186.00 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||
17833048
|
Brannan™
44/011/9 + 49/503/0 |
-20°C to +150°C | Spirit | Total Immersion | 30.5 cm |
184.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17852988
|
Brannan™
44/403/0 + 49/503/0 |
-10°C to +110°C | Red Spirit | Partial Immersion | 30.5 cm |
183.00 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||
17803048
|
Brannan™
44/430/5 + 49/503/0 |
-10°C to +110°C | Green Spirit | Total Immersion | 15 cm |
183.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17843048
|
Brannan™
44/406/0 + 49/503/0 |
-10°C to +150°C | Red Spirit | Partial Immersion | 30.5 cm |
186.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17813018
|
Brannan™
44/423/5 + 49/503/0 |
-10°C to +110°C | Green Spirit | Partial Immersion | 20.5 cm |
183.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17833058
|
Brannan™
44/401/1 + 49/503/0 |
-10°C to +50°C | Red Spirit | Total Immersion | 30.5 cm |
184.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17893048
|
Brannan™
44/401/0 + 49/503/0 |
-10°C to +50°C | Red Spirit | Partial Immersion | 30.5 cm |
186.00 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||
17823088
|
Brannan™
46/456/0 + 49/503/0 |
-100°C to +50°C | Red Spirit | Total Immersion | 30.5 cm |
293.00 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||
17803128
|
Brannan™
44/010/8 + 49/503/0 |
-20°C to +110°C | Spirit | Total Immersion | 30.5 cm |
183.00 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||
17813128
|
Brannan™
44/430/0 + 49/503/0 |
-10°C to +110°C | Red Spirit | Total Immersion | 15 cm |
183.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Lab thermometer
- Safe non-mercury filling
- Supplied with calibration certificate
- Non-roll end cap
- Supplied as standard in protective individual, resealable telescopic tube
- Made in UK
Specifications
ISO 17025 UKAS | |
Glass | |
1.2 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Brannan™ Lab Thermometer with 3-points UKAS Calibration