Learn More
Brannan™ Lab Thermometer with 3-points UKAS Calibration
General purpose laboratory thermometer complete with calibration certificate.
Brand:  Brannan™ 44/011/9 + 49/503/0
 View more versions of this product
Product Code. 17833048
  • 184.00 € / Each
Explore more special offers
Description
Description
  • Lab thermometer
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications
Specifications
0°C, 70°C, 140°C
Total Immersion
Glass
302°F
-4°F
-4°F to +302°F
± 1.0°C at 35°C
1.2 cm
ISO 17025 UKAS
30.5 cm
Spirit
150°C
-20°C
-20°C to +150°C
1°C, 2°C