Learn More
Jenway™ Genova-Series Spectrophotometer Accessory, Microcuvette Holder
Parts and accessories to expand analysis capabilities or make testing easier.
Brand:  Jenway™ 83056-06
Product Code. 15717567
  • 228.00 € / Each
Explore more special offers
Description
Description
  • Accessories
  • For use with Genova-Series Spectrophotometer
Specifications
Specifications
10 x 10 mm square, 8.5 mm beam height
Microcuvette Holder
Genova-Series Spectrophotometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Jenway™ Genova-Series Spectrophotometer Accessory, Microcuvette Holder > Microcuvette Holder