missing translation for 'onlineSavingsMsg'
Learn More

Brady™ GHS Symbol - Flammable, Individual Markers on a Roll

Pre-printed labels to identify your chemical product containers.

Brand:  Brady™ 834186

71.79 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 15614837

  • 84.18 € / Pack of 250
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications

Specifications

PIC 1802-21*21-B7541-RLL-DC
Polyester
Permanent Acrylic
0.099 mm
-40°C to 110°C
Black on White
Gloss
250/Pk.
21 mm
21 mm
Diamond
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ GHS Symbol - Flammable, Individual Markers on a Roll > W x H: 21 x 21 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.