Learn More
Gibco™ DPBS (10X), no calcium, no magnesium
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Brand: Gibco™ 14200075
59.10 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.
Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).
We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.
This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature
Specifications
No Sodium Pyruvate | |
Clear | |
10 X | |
DPBS | |
Dulbecco's Phosphate Buffered Saline | |
2630 - 3000 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Animal Origin-Free | |
Storage conditions: 15°C to 30°C Shipping conditions: Room temperature Shelf life: 36 months from date of manufacture |
|
Gibco™ | |
500 mL | |
Mammalian Cell Culture | |
6.7 - 7.0 | |
Room Temperature | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.