missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS (10X), no calcium, no magnesium
GREENER_CHOICE

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

65.25 €

Specifications

Additive No Sodium Pyruvate
Classification Animal Origin-Free
Color Clear
Content And Storage Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture
Dilution 10 X
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15217168
GREENER_CHOICE
View Documents
Gibco™
14200075
500 mL
65.25 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).

We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

No Sodium Pyruvate
Clear
10 X
DPBS
Mammalian Cell Culture
2630 - 3000 mOsm/kg
No Phenol Red
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Animal Origin-Free
Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture
Gibco™
Dulbecco's Phosphate Buffered Saline
Sustainable packaging
6.7 - 7.0
Room Temperature
Liquid
Videos
SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.