missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate), Powder

Produced by bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells

Brand:  Gibco™ 11811098

2052.00 EUR valid until 2025-12-31
Use promo code "24361" to get your promotional price.


 View more versions of this product

Product Code. 11548676

  • 2275.00 € / 25g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Resistance to Geneticin™ Selective Antibiotic (G418 Sulfate) is conferred by the E. coli APH (3) – I and APH (3) – II resistance genes.

  • Used as selective antibiotic in concentration range of 100 - 200μg/mL for bacteria, or 200 – 500μg/mL for most mammalian cells
  • Supplied as powder and should be made into a stock solution of 10 – 50mg/mL in water

Cell Culture, Cloning, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection, Transformation

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
10 to 50 μg/mL
Powder
Sterile
Geneticin™
Room Temperature
Eukaryotic Cells, Prokaryotic Cells
Mammalian Cell Culture, Insect Cell Culture
Antibiotic
Eukaryotic Selection⁄Stable Cell Line Generation
25 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.