missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ HEPES (1M)
A zwitterionic organic chemical buffering agent commonly used in cell culture media
Brand: Gibco™ 15630080
119.70 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.
This item is not returnable.
View return policy
Description
- HEPES (N-2-hydroxyethylpiperazine-N-2-ethane sulfonic acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media
- The addition of 10–25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator
- A good buffering choice for many cell culture systems because it is membrane impermeable, has limited effect on biochemical reactions, is chemically and enzymatically stable
- It also has very low visible and UV light absorbance
- Store at 2°C to 8°C
Order Info
Shipping Condition: Room Temperature
Specifications
Zwitterionic Organic Chemical Buffer | |
Chromatin Biology | |
100 mL | |
N-2-Hydroxyethylpiperazine-N-2-Ethane Sulfonic Acid | |
Sterile-filtered |
Clear | |
Liquid | |
Buffer | |
Sterile | |
Biochemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Spot an opportunity for improvement?Share a Content Correction