Learn More
Gibco™ HEPES (1M)

A zwitterionic organic chemical buffering agent commonly used in cell culture media
135.00 €
Specifications
Chemical Name or Material | Zwitterionic Organic Chemical Buffer |
---|---|
For Use With (Application) | Chromatin Biology |
Physical Form | Liquid |
Solution Type | Buffer |
Sterility | Sterile |
Description
- HEPES (N-2-hydroxyethylpiperazine-N-2-ethane sulfonic acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media
- The addition of 10–25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator
- A good buffering choice for many cell culture systems because it is membrane impermeable, has limited effect on biochemical reactions, is chemically and enzymatically stable
- It also has very low visible and UV light absorbance
- Store at 2°C to 8°C
Order Info
Shipping Condition: Room Temperature
Specifications
Zwitterionic Organic Chemical Buffer | |
Liquid | |
Sterile | |
Biochemical |
Chromatin Biology | |
Buffer | |
Sterile-filtered |
Safety and Handling
Recommended Storage : Storage conditions: 2°C to 8°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.