missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Hybridoma-SFM, powder

Complete, ready-to-use formulation

Brand:  Gibco™ 12300067

284.75 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 11520426

  • 339.00 € / 10 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Gibco Hybridoma SFM is a serum-free, very low-protein medium specifically developed for the ability to support the growth of hybridomas for antibody production.

  • Ability to support suspension and stationary hybridomas and myelomas
  • Serum-free, very low-protein formulation
  • Supports suspension and stationary hybridomas and myelomas
  • Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas
  • Provides high cell densities and IgG production
  • Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
  • Complete, ready-to-use formulation
  • No supplementation is required
  • Serum-free, very low-protein formulation, allowing for easier purification of your protein of interest
  • Contains < 20μg/mL total protein

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8°C
Shipping conditions: Ambient
Shelf life: 33 months from date of manufacture
Low Protein, Serum-free
Hybridoma Medium, powder
Glutamine
10 L
Hybridomas
Powder
Serum-free
Gibco™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2558703 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12300067
Lot Number2558704 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12300067
Lot Number2553364 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12300067
Lot Number2522388 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12300067
Lot Number2512450 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12300067
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Hybridoma-SFM, powder > 10L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.