Learn More
Gibco™ Opti-MEM™ I Reduced Serum Medium, no phenol red

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in cell growth rate or morphology.
Brand: Gibco™ 11058021
72.26 EUR valid until 2025-12-31
Use promo code "24273" to get your promotional price.
Description
Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in cell growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I Reduced-Serum Medium modifications for a range of cell culture applications.
This Opti-MEM™ is modified as follows:
With: L-glutamine
Without: Phenol Red
The complete formulation is confidential. For more information, please contact Technical Services.
Using Opti-MEM™ Reduced-Serum Medium
Opti-MEM™ I medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature

Specifications
Sp2, AE-1, CHO, BHK-21, and HEK | |
Human Origin | |
Liquid | |
Sterile-filtered | |
No Phenol Red | |
Gibco™, Opti-MEM™ | |
Room Temperature |
Primary Fibroblasts | |
1 X | |
Opti-MEM I | |
Glutamine | |
cGMP-compliant under the ISO 13485 standard | |
500 mL | |
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2670831 | Certificate of Analysis | 29/01/2025 | 11058021 |
2660204 | Certificate of Analysis | 29/01/2025 | 11058021 |
2715536 | Certificate of Analysis | 29/01/2025 | 11058021 |
2813963 | Certificate of Analysis | 28/01/2025 | 11058021 |
2731457 | Certificate of Analysis | 28/01/2025 | 11058021 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.