missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Penicillin-Streptomycin (5,000 U/mL)

For prevention from bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.

Brand:  Gibco™ 15070063


Product Code. 11528876

  • 25.48 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Penicillin was originally purified from fungi Penicillium and acts by interfering directly with turnover of bacterial cell wall and indirectly by triggering release of enzymes that alter cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to 30S subunit of the bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria.

  • Solution contains 5000 units/mL of penicillin and 5000μg/mL of streptomycin

Cell Culture, Mammalian Cell Culture, Primary Cell Culture, T Cell Culture

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: -5°C to -20°C
Shipping conditions: Dry ice
Shelf life: 12 months from date of manufacture
Mammalian Cell Culture, Insect Cell Culture
Antibiotic
Prevention of Cell Culture Contamination
Dry Ice
100 X
Liquid
Sterile-filtered
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • PENICILLIN STREPTOMYCIN SOL

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P271-Use only outdoors or in a well-ventilated area.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : 4-Thia-1-azabicyclo¢3.2.0!heptane-2-carboxylic acid, 3, 3-dimethyl-7-oxo-6-¢(phenylacetyl)amino!-¢2-

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.