missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PFHM-II Protein-Free Hybridoma Medium (liquid)
GREENER_CHOICE

Serum-free, protein-free medium specifically developed for the ability to support the growth of hybridomas for antibody production

Brand:  Gibco™ 12040077

Product Code. 11370882

  • 117.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ability to support suspension and stationary hybridomas and myelomas
  • Complete, ready-to-use, serum-free, protein-free formulation
  • Ability to support suspension and stationary hybridomas and myelomas
  • Suitable for culture of recombinant myeloma lines as well as traditional hybridomas, Gibco PFHM II provides high cell densities and IgG production
  • Cholesterol-dependent cultures such as NS0 require the addition of 250x Cholesterol Lipid Concentrate
  • Sustainable packaging

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Protein-free
Liquid
Serum-free
Gibco™
Room Temperature
Hybridomas
Suspension Cell Culture
Hybridoma Medium
Glutamine, Phenol Red
1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2559742 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12040077
Lot Number2517641 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12040077
Lot Number2517643 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12040077
Lot Number2517649 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number12040077
Lot Number2824458 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number12040077
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ PFHM-II Protein-Free Hybridoma Medium (liquid) > Quantity: 1000 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.