Learn More
Gibco™ Trypsin (0.25%), phenol red

Made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas
30.61 €
Specifications
Chelators | No EDTA |
---|---|
Content And Storage | Storage conditions: -5 to -20°C. Shelf life: 24 months from date of manufacture. |
Cell Type | Mammalian Cells |
Classification | Animal Origin |
Concentration | 1 X |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12579069
![]() |
Gibco™
15050065 |
100 mL |
30.61 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Gibco™ Trypsin solutions are made from trypsin powder—an irradiated mixture of proteases derived from porcine pancreas
- Due to its digestive strength it is widely used for cell dissociation, during routine cell culture passaging and primary tissue dissociation
- This solution is formulated with phenol red indicator, and without EDTA
- Gibco™ Trypsin solutions are tested for pH, osmolality, sterility and performance
- Prior to manufacturing the raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal and viral contamination, as well as multiple activity assays, ash analysis and moisture analysis
- Lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency are all available
- Solution should be stored at -5°C to -20°C
Order Info
Shipping Condition: Dry Ice
Compliance
Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards.
Specifications
No EDTA | |
Mammalian Cells | |
1 X | |
Cell Culture Dissociation Reagent | |
Sterile-filtered | |
280 - 310 mOsm/kg | |
Wet Ice | |
In vitro Bioassay |
Storage conditions: -5 to -20°C. Shelf life: 24 months from date of manufacture. |
|
Animal Origin | |
Liquid | |
Sterile-filtered | |
Phenol Red | |
Gibco™ | |
Animal Origin |
Safety and Handling
missing translation for 'shelfLife' : 24 Months
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.