missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Trypsin (0.25%), phenol red
GREENER_CHOICE

Made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas

Brand:  Gibco™ 15050065

 View more versions of this product

Product Code. 12579069

  • 30.61 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Gibco™ Trypsin solutions are made from trypsin powder—an irradiated mixture of proteases derived from porcine pancreas
  • Due to its digestive strength it is widely used for cell dissociation, during routine cell culture passaging and primary tissue dissociation
  • This solution is formulated with phenol red indicator, and without EDTA
  • Gibco™ Trypsin solutions are tested for pH, osmolality, sterility and performance
  • Prior to manufacturing the raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal and viral contamination, as well as multiple activity assays, ash analysis and moisture analysis
  • Lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency are all available
  • Solution should be stored at -5°C to -20°C

Order Info

Shipping Condition: Dry Ice

Compliance

Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No EDTA
Mammalian Cells
1 X
Cell Culture Dissociation Reagent
Sterile-filtered
280 - 310 mOsm/kg
100 mL
Animal Origin
Storage conditions: -5 to -20°C.
Shelf life: 24 months from date of manufacture.
Animal Origin
Liquid
Sterile-filtered
Phenol Red
Gibco™
Wet Ice
In vitro Bioassay
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2500304 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15050065
Lot Number2679067 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15050065
Lot Number2679069 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15050065
Lot Number2500314 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15050065
Lot Number2753822 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15050065
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Trypsin (0.25%), phenol red > Quantity: 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.