missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jenway™ Glass Bench pH Meter
Versatile, easy-to-use pH, mV, and temperature meter. Jenway™ Glass Bench pH Meter is ideal for routine analysis.
Brand: Jenway™ 35459-06
695.00 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- Simultaneous readout of pH and temperature
- pH resolution to 3 decimal places
- 1, 2 or 3 point calibration
- Automatic or manual buffer selection
- Storage of up to 32 results
- RS232 connection to printer or PC via DataWay
- 3 year warranty
Specifications
Glass combination pH electrode (924 005), electrode stand and holder, ATC probe (027 500), BNC shorting plug, pH 4, 7 and 10 buffers and EU power supply | |
±0.2 | |
Analog, RS-232 | |
0.001, 0.01, 0.1 | |
1 to 3 | |
0.1°C | |
850 g | |
EU Power Supply | |
Bench pH Meter | |
250 mm | |
9 V |
±0.5°C | |
±0.003 | |
-2.000 to 19.999 | |
0.1/1mV | |
-10°C to +105°C | |
LCD | |
3 Years | |
ATC | |
210 mm | |
55 mm | |
50/60 Hz |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Jenway™ Glass Bench pH Meter > Includes: Glass combination pH electrode (924 005), electrode stand and holder, ATC probe (027 500), BNC shorting plug, pH 4, 7 and 10 buffers and EU power supply
Spot an opportunity for improvement?Share a Content Correction