Learn More
DWK Life Sciences Kimble™ Glass-filled Nylon Connectors with PTFE Seals
Kimble™ Glass-filled Nylon Connectors with PTFE Seals are threaded connectors used for coupling two items with a GPI finish.
26.90 € - 26.95 €
Specifications
Material Nylon
Product Type Connector
For Use With (Application) To connect standard rotary evaporator traps to vials
Includes PTFE seals
View More Specs
Products 2
Product Code Brand Thread Size Price Quantity & Availability  
Product Code Brand Thread Size Price Quantity & Availability  
17751088
View Documents
DWK Life Sciences
747205-1320
13-425 to 20-400
26.95 €
Each
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
15389796
View Documents
DWK Life Sciences
747205-1315
13-425 to 15-425
26.90 €
Each
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
Description
Description
  • Connectors have PTFE seals and FKM O-Rings
  • Blue glass-filled nylon
  • For use to 150°C
Specifications
Specifications
Nylon
Connector
To connect standard rotary evaporator traps to vials
PTFE seals
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Glass-filled Nylon Connectors with PTFE Seals