missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Glass-filled Nylon Connectors with PTFE Seals

Kimble™ Glass-filled Nylon Connectors with PTFE Seals are threaded connectors used for coupling two items with a GPI finish.

Brand:  DWK Life Sciences 747205-1315

 View more versions of this product

Product Code. 15389796

  • 26.90 € / Each
Estimated Shipment: 15-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Connectors have PTFE seals and FKM O-Rings
  • Blue glass-filled nylon
  • For use to 150°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

To connect standard rotary evaporator traps to vials
13-425 to 15-425
Connector
PTFE seals
Nylon
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Kimble™ Glass-filled Nylon Connectors with PTFE Seals > Connector; 13mm-425 to 15mm-425

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.