Learn More
Macherey-Nagel™ Glass Flash Chromatography Column
Brand:  Macherey-Nagel™ 727404
 View more versions of this product
Product Code. 10164801
  • 321.00 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
300 mm
I.D.: 30 mm
with Adapter and PTFE Tap
Suited The Separation of Compounds up to Gram Levels No Expensive Equipment Required
Glass
Flash Column
Flash Chromatography Glass Column