missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Soda Lime Glass Pasteur Pipette
Soda Lime Glass Pasteur Pipette is intended for single use in microbiological and serological laboratories.
73.84 € - 150.00 €
Specifications
Autoclavable | Not Autoclavable |
---|---|
Material | Soda Lime Glass |
For Use With (Application) | Single Use in Microbiological and Serological Laboratory Applications, Pipetting Liquids, Dropwise Dosing of Liquids and Reagents, Distribution of Heated Liquid Quantities |
Quantity | 1000 Pk. |
Volume (English) | 0.07 oz. |
Product Code | Brand | Length (Metric) | Type | Tip Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Type | Tip Style | Price | Quantity & Availability | ||||
11546963
|
Fisherbrand™
M4150NO250FS4 |
150 mm | Unplugged Pasteur Pipet | Standard Tip |
93.60 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||
11506973
|
Fisherbrand™
M4150NW250FS4 |
150 mm | Cotton-plugged Pasteur Pipet | Standard Tip |
124.00 €
Pack of 1000 |
In Stock
Log in to see stock. |
||||
11566963
|
Fisherbrand™
M4230NO250FS4 |
230 mm | Unplugged Pasteur Pipet | Standard Tip |
96.60 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||
11765098
|
Fisherbrand™
M4230NW250FS4 |
230 mm | Cotton-plugged Pasteur Pipet | Standard Tip |
126.00 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||
11755108
|
Fisherbrand™
M4270NO250FS4 |
270 mm | Unplugged Pasteur Pipet | Standard Tip |
115.00 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||
11795098
|
Fisherbrand™
M4270NW250FS4 |
270 mm | Cotton-plugged Pasteur Pipet | Standard Tip |
150.00 €
Pack of 1000 |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||
11775098
|
Fisherbrand™
M4230OO250FS4 |
230 mm | Unplugged Pasteur Pipet | Closed Tip |
73.84 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||
11785098
|
Fisherbrand™
M4230OW250FS4 |
230 mm | Cotton-plugged Pasteur Pipet | Closed Tip |
107.00 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||
Description
- Pasteur pipettes made of soda-lime glass are a good alternative to plastic pipettes
- Complies with ISO 7712
- Pasteur pipettes with short or long extended, fine tip (150 mm, 230 mm and 270 mm)
- Pasteur pipettes with and without cotton plug
- The filling volume is approx. 2 mL
- Ideal for use with Fisherbrand Pipettors
Specifications
Not Autoclavable | |
Single Use in Microbiological and Serological Laboratory Applications, Pipetting Liquids, Dropwise Dosing of Liquids and Reagents, Distribution of Heated Liquid Quantities | |
0.07 oz. | |
Single-use |
Soda Lime Glass | |
1000 Pk. | |
2 mL | |
Fisherbrand Pipetters |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Soda Lime Glass Pasteur Pipette