Learn More
Macherey-Nagel™ Glass Plate Nano-Adamant UV254
Brand:  Macherey-Nagel™ 821120
 View more versions of this product
Product Code. 10661553
  • 457.00 € / Pack of 50
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
2 to 10 μm
50 Pk.
UV254
∼500 m2/g
10 x 20 cm
60 Å
Glass Plate
0.75 mL/g
0.20 mm